Vacuum pumps, Rotavac Valve Control
Supplier: Heidolph
531-0376EA 1909 EUR
531-0376 531-3693 HEID591-00130-04
Vacuum pumps, Rotavac Valve Control
Pumps Vacuum Rotary Evaporator Pumps
Rotavac Valve Control. Two stage diaphragm pump made from chemically resistant materials. Suction capacity for use with up to three rotary evaporators at the same time.
  • High suction capacity of 1,7 m³/h, for fastest evacuation
  • Ultimate vacuum of 9 mbar
Can be combined with a condensate cooler.
Designed for all Hei-VAP rotary evaporators.
Informace pro objednávání: Other vacuum pump options are available, please contact Avantor for details.