VWR® Aquaresist Water Bath Protective Agent
Supplier: Avantor
VWR®
462-7051EA 31.5 EUR
462-7051 462-0600 462-7000
VWR® Aquaresist Water Bath Protective Agent
Cleaning Agents Water Bath Protection Reagents
Prevents the growth of algae and bacteria in baths and circulating thermostats, for optimally hygienic working conditions.
  • Durable: Effective for weeks, as displayed by a colour indicator
  • Economical: Low consumption - only 1 ml solution per 1 litre of water
  • Available with and without colour indicator
Upozornení: Use biocides safely. Always read the label and product information before use.