Ultrapure water systems, Simplicity®
Supplier: Merck
SIMSV00SZ SIMSV00FR SIMS000UK SIMSV00UK SIMS000BE SIMS000GE SIMS000FR SIMSV00BE SIMSV00SP SIMS000SZ SIMS000EU SIMSV00EU SIMS000IT SIMSV00IT SIMS000SP SIMSV00GE
171-0306EA 5560 EUR
171-0306 171-0765 171-0275 171-0749 171-0762 171-0298 171-0748 171-0764 171-0305 171-0301 171-0297 171-0302 171-0299 171-0304 171-0300 171-0303
Ultrapure water systems, Simplicity®
Water Purification Systems Pure and Ultra Pure Water Systems
The Simplicity® portable water purification system can be located anywhere in your laboratory. It requires no special installation, no water feed connection and produces ultrapure water on demand from pretreated water (Elix® system, distilled, deionised or reverse osmosis water). The Simplicity® system is an economical alternative for laboratories that currently use bottled water. Simplicity® ultrapure water, produced and used at the point-of-use, is not subject to the degradation and contamination that affects bottled water. The intuitive colour graphic display of the Simplicity® system means that the user can instantly see all system parameters, including water quality, reservoir level and whether the cartridge needs replacing. Type I ultrapure water for scientists who require less than 5 litres per day.
  • During breaks in collection, constant water quality is guaranteed  by automatic recirculation
  • Easy installation: The user simply connects this to an electrical supply, connect the SimpliPak® cartridge, and then fill the removable reservoir with pre-treated water
  • User friendly display
  • System draws water from a removable, refillable, built-in reservoir
  • Maintenance reduced to a single cartridge change once or twice a year, based on the resistivity value displayed for the ultrapure water produced; treatment module and filter can be changed without tools
The Simplicity® UV system contains a built-in dual wavelength UV lamp (185 and 254 nm) to reduce organic contaminants to <5 ppb making the water suitable for sensitive applications such as HPLC, GC and TOC analysis.
Power supply: 100 to 230 V/50 to 60 Hz.
Power supply: 100 - 230 V - 50 - 60 Hz
Informace pro objednávání: SimpliPak® should be added with a Millipak® or BioPak® final filter.
Upozornení: * with BioPak® ultrafiltration cartridge as final polisher.