Ultrapure water systems, Synergy®
Supplier: Merck
Synergy®
172-5160EA 5590 EUR
172-5160 172-5159 171-0712 171-0699 171-0314 171-0711 171-0697 171-0774 171-0312 171-0698 171-0313 171-0772 171-0310 171-0695 171-0311 171-0696 171-0773 171-0770 171-0693 171-0771 171-0694 171-0691 171-0692 171-0309 171-0307 171-0308 171-0767 171-0768 171-0688 171-0689 171-0686 171-0763 171-0687 171-0684 171-0685 171-0088
Ultrapure water systems, Synergy®
Water Purification Systems Pure and Ultra Pure Water Systems
Synergy® ultrapure water systems are ideal for laboratories with access to purified water that needs a 'point of use' supply of high quality ultrapure water. The Synergy® system is suitable for users needing 2 to 20 L/day. The system produces Type I water from pre-treated water at a flow from 1 to 1,5 L/min. The water produced is suitable for the preparation of buffers for liquid chromatography, biochemical experiments, isocratic or gradient solutions for HPLC analysis, culture media for bacteriological analysis or cell cultures and solutions for spectrophotometry, spectroscopy and other analytical techniques.
  • Can be operated with or without remote dispenser
  • Easy monitoring: measures and displays water quality on its two-line, alphanumeric display with a choice of six languages
  • Automatic recirculation allows the system to maintain water quality in-between periods of use
  • Simple to operate and maintain: Unique purification packs simplify cartridge replacement
  • Built-in dispense timer simplifies the dispensing operation and reduces operator attendance
  • UV photo-oxidation for low organic contamination (UV model only)
  • The system's compact design, along with the versatile remote dispenser, ensures easy installation anywhere in the laboratory, either on the bench or wall mounted
The system monitor informs the user when cartridge exchange is required or when the UV lamp needs replacing (UV model only). A built-in timer allows the operator to deliver water for a fixed time period. The system automatically stops once the time period has elapsed. The time period can easily be set by the user between 1 and 60 minutes and, remains stored in the memory until modified.
* With BioPak® final.
Informace pro objednávání: SynergyPak® cartridge should be added with a Millipak® or BioPak® final filter.