(+)-Cinchonine, Sigma-Aldrich®

Supplier: Merck

Synonyms: Cinchonine, (S)-Quinolin-4-yl((1S,2R,4S,5R)-5-vinylquinuclidin-2-yl)methanol

27370-100G 27370-25G
SIAL27370-100GEA 153 EUR
SIAL27370-100G SIAL27370-25G
(+)-Cinchonine, Sigma-Aldrich®
(+)-Cinchonine
Formula: C₁₉H₂₂N₂O
MW: 294.4 g/mol
Melting Pt: 261…264 °C
Density: 1.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064372
CAS Number: 118-10-5
EINECS: 204-234-6
Merck Index: 13,02308

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR