(-)-Quinine, Sigma-Aldrich®
Supplier: Merck
Synonyms: (R)-[(2S,4S,5R)-1-Aza-5-vinylbicyclo[2.2.2]oct-2-yl](6-methoxyquinolin-4-yl)methanol, Quinine, (R)-(6-methoxyquinolin-4-yl)((1S,2S,4S,5R)-5-vinylquinuclidin-2-yl)methanol
M-Clarity™ quality level = MQ200
145904-10G 145904-50G
SIAL145904-10GEA 66.4 EUR
SIAL145904-10G SIAL145904-50G
(-)-Quinine, Sigma-Aldrich®
(-)-Quinine
Formula: C₂₀H₂₄N₂O₂
Melting Pt: 176 °C
MDL Number: MFCD00198096
CAS Number: 130-95-0
EINECS: 205-003-2
Merck Index: 12,08245