Antimony(III) potassium oxitartrate trihydrate 99+%
Supplier: Thermo Scientific
Warning
Synonyms: (R,R)-(+)-Antimony potassium tartrate trihydrate, (+)-Antimony potassium L-tartrate trihydrate, Tartar emetic, Antimony potassium L(+)-tartrate trihydrate, L(+)-Tartaric acid antimony potassium salt trihydrate, Potassium Antimony (III) Oxide Tartrate Trihydrate, (R,R)-(+)-Tartaric acid antimony potassium salt trihydrate, L(+)-Antimony potassium tartrate trihydrate, Potassium antimony(III) oxide tartrate trihydrate, Antimony potassium (+) tartrate trihydrate, Potassium antimony tartrate trihydrate, Potassium antimonyl tartrate sesquihydrate, Antimony potassium tartrate trihydrate, Potassium antimonyl tartrate trihydrate
223801000 223805000 223800025
ACRO223801000EA 41.6 EUR
ACRO223801000 ACRO223805000 ACRO223800025
Antimony(III) potassium oxitartrate trihydrate 99+%
Antimony (III) potassium oxitartrate trihydrate
Formula: C₈H₄K₂O₁₂Sb₂·3H₂O
MW: 667.88 g/mol
Melting Pt: 100 °C
Density: 2,6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00167056
CAS Number: 28300-74-5
EINECS: 229-436-1
UN: 1551
ADR: 6.1,III
Merck Index: 13,00706
Order Now
Specification Test Results
Appearance White Crystalline powder
Infrared spectrum Conforms
Titration Iodometric ≥99.0 %
Loss on drying ≤2.7 % (105°C)