X-GlcA ((5-bromo-4-chloro-3-indolyl-ß-D-glucuronide) cyclohexylammonium salt), Sigma-Aldrich®
Supplier: Merck
Synonyms: X-Gluc
M-Clarity™ quality level = MQ200
B6650-100MG B6650-10MG B6650-25MG
SIALB6650-100MGEA 647 EUR
SIALB6650-100MG SIALB6650-10MG SIALB6650-25MG
X-GlcA ((5-bromo-4-chloro-3-indolyl-ß-D-glucuronide) cyclohexylammonium salt), Sigma-Aldrich®
X-GlcA ((5-Bromo-4-chloro-3-indolyl-ß-D-glucuronide) cyclohexylammonium salt)
Formula: C₂₀H₂₆BrClN₂O₇
MW: 521.79 g/mol
Melting Pt: 230 °C
Storage Temperature: Freezer
MDL Number: MFCD00058543
CAS Number: 114162-64-0