Dextrin, precipitated_alcohol
Supplier: Thermo Fisher Scientific
Synonyms: Dextrin
A15717.36 A15717.22
A15717.36EA 101 EUR
A15717.36 A15717.22
Dextrin, precipitated_alcohol
Dextrin
CAS No.: 9004-53-9 Merck Reference: 14,2953 MDL No.: MFCD00081554
Formula: (C₆H₁₂O₆)n
MW: 180.16 g/mol
Density: 1.45 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00081554
CAS Number: 9004-53-9
EINECS: 232-675-4
Merck Index: 14,2953