Iron(II) sulphate hydrate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Ferrous sulphate hydrate
M-Clarity™ quality level = MQ100
450278-25G 450278-5G
SIAL450278-25GEA 364 EUR
SIAL450278-25G SIAL450278-5G
Iron(II) sulphate hydrate, Sigma-Aldrich®
Iron (II) sulphate hydrate
Formula: FeSO₄·H₂O
Melting Pt: ∼300 °C
Density: 2.97 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149722
CAS Number: 13463-43-9
EINECS: 231-753-5