May Grunwald's eosin-methylene blue solution
Supplier: Roth Carl
Danger
T863.1 T863.2 T863.3
ROTHT863.1EA 35 EUR
ROTHT863.1 ROTHT863.2 ROTHT863.3
May Grunwald's eosin-methylene blue solution
May Grunwald's eosin-methylene blue
Storage Temperature: Ambient
MDL Number: MFCD00131580
UN: 1230
ADR: 3,II