Litmus

Supplier: Roth Carl

0248.1 0248.2
ROTH0248.1EA 17.8 EUR
ROTH0248.1 ROTH0248.2
Litmus
Litmus
MDL Number: MFCD00131528
CAS Number: 1393-92-6
EINECS: 215-739-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR