β-Nicotinamide adenine dinucleotid, oxidised form (NAD, oxidised form), Sigma-Aldrich®

Supplier: ROCHE


M-Clarity™ quality level = MQ100

ROCH10621650001EA 560 EUR
ROCH10621650001 ROCH621650
β-Nicotinamide adenine dinucleotid, oxidised form (NAD, oxidised form), Sigma-Aldrich®
β-Nicotinamide adenine dinucleotid, oxidised form (NAD, oxidized form)
Formula: C₂₁H₂₇N₇O₁₄P₂
MW: 663,43 g/mol
Melting Pt: 140 to 142 °C
MDL Number: MFCD00036253
CAS Number: 53-84-9
Merck Index: 12,06429

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR