β-Nicotinamide adenine dinucleotid, oxidised form (NAD, oxidised form)
Supplier: Roth Carl
Synonyms: Nadide, NAD+, Nicotinamide adenine dinucleotide, oxidized
AE11.1 AE11.2 AE11.3
ROTHAE11.1EA 38.6 EUR
ROTHAE11.1 ROTHAE11.2 ROTHAE11.3
β-Nicotinamide adenine dinucleotid, oxidised form (NAD, oxidised form)
β-Nicotinamide adenine dinucleotide (NAD, oxidized form)
Formula: C₂₁H₂₇N₇O₁₄P₂
MW: 663.43 g/mol
Melting Pt: 140 to 142 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00036253
CAS Number: 53-84-9
EINECS: 200-184-4
Merck Index: 13,06370