ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form)
Supplier: ROCHE
M-Clarity™ quality level = MQ100
ROCH10240354001EA 2170 EUR
ROCH10240354001 ROCH240354
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form)
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form)
Formula: C₂₁H₂₆N₇Na₂O₁₇P₃
MW: 787,37 g/mol
MDL Number: MFCD00065390
CAS Number: 24292-60-2