ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form)

Supplier: ROCHE


M-Clarity™ quality level = MQ100

ROCH10128058001EA 876 EUR
ROCH10128058001 ROCH128058
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form)
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form)
Formula: C₂₁H₂₆N₇Na₂O₁₇P₃
MW: 787,37 g/mol
MDL Number: MFCD00065390
CAS Number: 24292-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR