Zinc carbonate basic (>58.0% Zn) ≥97%
Supplier: Thermo Fisher Scientific
Synonyms: Dicarbonatohexahydroxypentazinc, Zinc carbonate hydroxide, Zinc hydroxide carbonate
A14590.0B A14590.30
A14590.0BEA 66.6 EUR
A14590.0B A14590.30
Zinc carbonate basic (>58.0% Zn) ≥97%
Zinc carbonate basic
Purity: 97% CAS No.: 5263-02-5 Molecular Formula: Zn5(CO3)2(OH)6 Formula Weight: 548.97 MDL No.: MFCD00151660
Formula: C₂H₆O₁₂Zn₅
MW: 549.01 g/mol
Melting Pt: > 140 °C
Density: 3.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011519
CAS Number: 5263-02-5
EINECS: 226-076-7
UN: 3077
ADR: 9 III