(1S)-(-)-β-Pinene, Sigma-Aldrich®
Supplier: Merck
Synonyms: (-)-Pin-2(10)-ene
M-Clarity™ quality level = MQ100
402753-10G 402753-1G
SIAL402753-10GEA 202 EUR
SIAL402753-10G SIAL402753-1G
(1S)-(-)-β-Pinene, Sigma-Aldrich®
(1S)-(-)-β-Pinene
Formula: C₁₀H₁₆
MW: 136.24 g/mol
Boiling Pt: 165 °C (1013 hPa)
Melting Pt: –61 °C
Density: 0.9034 g/cm³ (20 °C)
Flash Pt: 36 °C
MDL Number: MFCD00001345
CAS Number: 18172-67-3
EINECS: 242-060-2
UN: 2319
ADR: 3,III
Merck Index: 12,07600