CAPS (3-(cyclohexylamino)propanesulphonic acid) 99% for biochemistry
Supplier: Thermo Scientific
Warning
Synonyms: 3-Cyclohexylaminopropanesulphonic acid
172621000 172620010 172622500
ACRO172621000EA 86.7 EUR
ACRO172621000 ACRO172620010 ACRO172622500
CAPS (3-(cyclohexylamino)propanesulphonic acid) 99% for biochemistry
CAPS (3-(Cyclohexylamino)propanesulphonic acid)
Formula: C₆H₁₁NH(CH₂)₃SO₃H
MW: 221.32 g/mol
Melting Pt: 324 °C (dec.)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00003837
CAS Number: 1135-40-6
EINECS: 214-492-1
Merck Index: 13,01773
Order Now
Specification Test Results
Appearance White Powder
Infrared spectrum Conforms
Loss on drying ≤5 %
UV at 280 nm A: ≤0.02
Titration with NaOH ≥98.5 % (On dry substance)