Nickel carbonate, anhydrous 98%
Supplier: Thermo Scientific
Danger
22897.18 22897.30
22897.18EA 51 EUR
22897.18 22897.30
Nickel carbonate, anhydrous 98%
Nickel carbonate
Purity: 98% CAS No.: 3333-67-3 Molecular Formula: CNiO3 Formula Weight: 118.72 Physical Form: -100 Mesh Powder Appearance: Pale purple UN No.: 3077 MDL No.: MFCD00011144
Formula: CNiO₃
MW: 118.7 g/mol
Density: 3.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011144
CAS Number: 3333-67-3
EINECS: 222-068-2
UN: 3077
ADR: 9,III