Capsaicin, Sigma-Aldrich®
Supplier: Merck
Synonyms:
8-Methyl-N-vanillyl-6-none-amid
M-Clarity™ quality level = MQ200
M2028-50MG
M2028-1G
M2028-250MG
SIALM2028-50MGEA
127
EUR
SIALM2028-50MG
SIALM2028-1G
SIALM2028-250MG
Capsaicin, Sigma-Aldrich®
Capsaicin
Formula:
C₁₈H₂₇NO₃ Boiling Pt: 210…220 °C (0.01 mmHg) Melting Pt: 66.1 °C |
MDL Number:
MFCD00017259 CAS Number: 404-86-4 EINECS: 206-969-8 UN: 2811 ADR: 6.1,III Merck Index: 12,01811 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...