Software for KingFisher™ systems, Bind-It™
Supplier: Thermo Scientific
733-1963EA 9620 EUR
FINN5189010
Software for KingFisher™ systems, Bind-It™
Bind-It™ software is designed to allow the user to develop custom made protocols or modify existing protocols for their own applications and run them in the KingFisher™ instruments. Additionally, Bind-It™ software enables user control access levels to be set, advanced protocol management, sample information and clear reporting. Predefined and validated purification protocols are available for different applications, offering the possibility to choose the best possible method for a sample process. The Bind-It™ software also enables the KingFisher™ Flex to interface with liquid handling, robotics and plate stacking instruments, providing a fully automated solution and the highest throughput.