VWR® Sharps Container Systems, Red
Supplier: Avantor
119-0902EA 133 EUR
119-0902 119-0901 119-0911 119-0900 119-0910 119-0094 119-0093 119-0091 119-0909 119-0908 119-0906 119-0905 119-0904 119-0903
VWR® Sharps Container Systems, Red
Bins Sharps Containers
Sharps container systems provide solutions for the safe and effective containment and disposal of contaminated needles, syringes, and other sharps. Choose from three different systems in a variety of sizes.
  • Puncture-resistant and autoclave- and incinerator-safe
  • Transparent, locking lids
  • Needle removal ports
The stackable sharps containers are puncture-resistant and autoclave- and incinerator-safe. They feature transparent, locking lids, and needle removal ports. The tortuous path systems restrict hand entry.
The extended neck sharps containers feature a funneled vertical drop and extended handles. Puncture-resistant and autoclave- and incinerator-safe.