Tubing, silicone, for perimatic GP peristaltic pump dispenser
Supplier: Jencons
JENC256-028EA 76.5 EUR
JENC256-028 JENC256-029 JENC256-027
Tubing, silicone, for perimatic GP peristaltic pump dispenser
Silicone tubing assemblies.
  • Very good chemical resistance
  • Very good temperature resistance
Upozornení: Tubing assembly consists of tubing, glass dispense nozzle, glass 'j' piece and glass sinker