Beryllium ≥99.4% (metals basis), foil, Thickness: 0.1 mm (0.004 in)
Supplier: Thermo Scientific
Danger
10105.FF 10105.FI
10105.FFEA 774 EUR
10105.FF 10105.FI
Beryllium ≥99.4% (metals basis), foil, Thickness: 0.1 mm (0.004 in)
Beryllium
Purity: 99.4% CAS No.: 7440-41-7 Physical Form: ≈0.12g/25x25mm, Vacuum tight Odour: Odourless MDL No.: MFCD00134032
Formula: Be
MW: 9.01 g/mol
Boiling Pt: 2970 °C (1013 hPa)
Melting Pt: 1278 °C
Density: 1.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134032
CAS Number: 7440-41-7
EINECS: 231-150-7
Merck Index: 12,01201