Beryllium ≥99.5% (metals basis), irregular lumps 3-38 mm (0.1-1.5 in)
Supplier: Thermo Scientific
Danger
12853.09 12853.18
12853.09EA 120 EUR
12853.09 12853.18
Beryllium ≥99.5% (metals basis), irregular lumps 3-38 mm (0.1-1.5 in)
Beryllium
Purity: 99.5% CAS No.: 7440-41-7 Odour: Odourless MDL No.: MFCD00134032
Formula: Be
MW: 9.01 g/mol
Boiling Pt: 2970 °C (1013 hPa)
Melting Pt: 1278 °C
Density: 1.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134032
CAS Number: 7440-41-7
EINECS: 231-150-7
Merck Index: 12,01201