Calcium ≥99.5% (metals basis), granules -6 mesh, redistilled
Supplier: Thermo Fisher Scientific
|
Danger
|
00875.30
00875.A1
00875.30EA
149
EUR
00875.30
00875.A1
Calcium ≥99.5% (metals basis), granules -6 mesh, redistilled
Calcium
Purity: 99.5%
CAS No.: 7440-70-2
Odour: Odourless
UN No.: 1401
MDL No.: MFCD00085314
Formula:
Ca Boiling Pt: 1484 °C (1013 hPa) Melting Pt: 850 °C Density: 1.54 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00085314 CAS Number: 7440-70-2 EINECS: 213-179-5 UN: 1401 ADR: 4.3,II Merck Index: 13,01644 |