Selenium ≥99.999% (metals basis), amorphous shot 1-6 mm (0.04-0.24 in)
Supplier: Thermo Scientific
Danger
00290.18 00290.30
00290.18EA 88.1 EUR
00290.18 00290.30
Selenium ≥99.999% (metals basis), amorphous shot 1-6 mm (0.04-0.24 in)
Selenium
Purity: 99.999+% CAS No.: 7782-49-2 Odour: Odourless MDL No.: MFCD00134090
Formula: Se
MW: 78.96 g/mol
Boiling Pt: ∼685 °C (1013 hPa)
Melting Pt: 217 °C
Density: 4.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134090
CAS Number: 7782-49-2
EINECS: 231-957-4
UN: 3283
ADR: 6.1,III
Merck Index: 12,08572