Selenium 99,999 Metals basis, powder -200 mesh
Supplier: Thermo Fisher Scientific
Danger
36208.18 36208.30
36208.18EA 258 EUR
36208.18 36208.30
Selenium 99,999 Metals basis, powder -200 mesh
Selenium
Purity: 99.999% CAS No.: 7782-49-2 UN No.: 3077 MDL No.: MFCD00134090
Formula: Se
MW: 78.96 g/mol
Boiling Pt: ∼685 °C (1013 hPa)
Melting Pt: 217
Density: 4.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134090
CAS Number: 7782-49-2
EINECS: 231-957-4
UN: 3288
ADR: 6.1 III
Merck Index: 12,08572