Tellurium ≥99.9999% (metals basis), lumps typ. 12 mm (0.47 in)
Supplier: Thermo Fisher Scientific
Danger
10758.18 10758.30
10758.18EA 283 EUR
10758.18 10758.30
Tellurium ≥99.9999% (metals basis), lumps typ. 12 mm (0.47 in)
Tellurium
Purity: 99.9999% CAS No.: 13494-80-9 Odour: Odourless MDL No.: MFCD00134062
Formula: Te
Boiling Pt: 990 °C (1013 hPa)
Melting Pt: 450 °C
Density: 8.83 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134062
CAS Number: 13494-80-9
EINECS: 236-813-4
UN: 3284
ADR: 6.1,III
Merck Index: 12,09273