Tellurium ≥99.99% (metals basis), broken ingot
Supplier: Thermo Scientific
Danger
11073.18 11073.30
11073.18EA 105 EUR
11073.18 11073.30
Tellurium ≥99.99% (metals basis), broken ingot
Tellurium
Purity: 99.99% CAS No.: 13494-80-9 Odour: Odourless MDL No.: MFCD00134062
Formula: Te
MW: 127.6 g/mol
Boiling Pt: 990 °C (1013 hPa)
Melting Pt: 450 °C
Density: 8.83 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134062
CAS Number: 13494-80-9
EINECS: 236-813-4
UN: 3284
ADR: 6.1,III
Merck Index: 12,09273