Tellurium ≥99.99% (metals basis), powder -30 mesh
Supplier: Thermo Fisher Scientific
Danger
11074.14 11074.22
11074.14EA 117 EUR
11074.14 11074.22
Tellurium ≥99.99% (metals basis), powder -30 mesh
Tellurium
Purity: 99.99% CAS No.: 13494-80-9 Odour: Odourless UN No.: 3288 MDL No.: MFCD00134062
Formula: Te
MW: 127.6 g/mol
Boiling Pt: 990
Melting Pt: 450
Density: 8.83 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134062
CAS Number: 13494-80-9
EINECS: 236-813-4
UN: 3288
ADR: 6.1 III
Merck Index: 12,09273