Copper 99,99 Metals basis, foil, annealed, Thickness: 0.127 mm (0.005 in)
Supplier: Thermo Fisher Scientific
00095.FIEA 129 EUR
00095.FI 00095.GB
Copper 99,99 Metals basis, foil, annealed, Thickness: 0.127 mm (0.005 in)
Copper
Purity: 99.99+% CAS No.: 7440-50-8 Physical Form: ≈2.8g/50x50mm Odour: Odourless MDL No.: MFCD00010965
Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545