Copper ≥99.9% (metals basis), shot 1-10 mm (0.04-0.4 in)
Supplier: Thermo Fisher Scientific
10159.36 10159.A3
10159.36EA 74.9 EUR
10159.36 10159.A3
Copper ≥99.9% (metals basis), shot 1-10 mm (0.04-0.4 in)
Copper
Purity: 99.9% CAS No.: 7440-50-8 Odour: Odourless MDL No.: MFCD00010965
Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545