Copper, gauze_wire 40 mesh, Ø 0.056 mm (0.0022 in)
Supplier: Thermo Fisher Scientific
40907.FL
40907.GT
40907.HB
40907.FLEA
162
EUR
40907.FL
40907.GT
40907.HB
Copper, gauze_wire 40 mesh, Ø 0.056 mm (0.0022 in)
Copper
CAS No.: 7440-50-8
Physical Form: Wire Cloth
Odour: Odourless
MDL No.: MFCD00010965
Formula:
Cu MW: 63.55 g/mol Boiling Pt: 2595 Melting Pt: 1083 Density: 8.94 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010965 CAS Number: 7440-50-8 EINECS: 231-159-6 Merck Index: 13,02545 |