Copper ≥99.9% (metals basis), spherical powder -170/+400 mesh oxygen 0.1%
Supplier: Thermo Fisher Scientific
Warning
43385.22 43385.A3 43385.36
43385.22EA 65.2 EUR
43385.22 43385.A3 43385.36
Copper ≥99.9% (metals basis), spherical powder -170/+400 mesh oxygen 0.1%
Copper
Purity: 99.9% CAS No.: 7440-50-8 Appearance: Copper Odour: Odourless UN No.: 3089 MDL No.: MFCD00010965
Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
UN: 3089
ADR: 4.1 III
Merck Index: 13,02545