Copper ≥99.999% (metals basis), plate, irregular, Puratronic® ∼ 10x34x0.6 cm (4x13.5x0.25 in)
Supplier: Thermo Scientific
43822.KSEA 808 EUR
43822.KS
Copper ≥99.999% (metals basis), plate, irregular, Puratronic® ∼ 10x34x0.6 cm (4x13.5x0.25 in)
Copper
Formula: Cu
MW: 63,55 g/mol
Boiling Pt: 2595 °C (1013 hPa)
Melting Pt: 1083 °C
Density: 8,94 g/cm³ (20 °C)
MDL Number: MFCD00010965
CAS Number: 7440-50-8
Merck Index: 13,02545