Copper ≥99.5% (metals basis), shot 0.8-2 mm (0.03-0.08 in)
Supplier: Thermo Fisher Scientific
44213.36 44213.A3 44213.22
44213.36EA 115 EUR
44213.36 44213.A3 44213.22
Copper ≥99.5% (metals basis), shot 0.8-2 mm (0.03-0.08 in)
Copper
Purity: 99.5% CAS No.: 7440-50-8 Odour: Odourless MDL No.: MFCD00010965
Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545