Copper ≥99.999% (metals basis), wire, Puratronic®, Ø 1.5 mm (0.06 in)
Supplier: Thermo Fisher Scientific
44550.G1 44550.G5
44550.G1EA 152 EUR
44550.G1 44550.G5
Copper ≥99.999% (metals basis), wire, Puratronic®, Ø 1.5 mm (0.06 in)
Copper
Purity: 99.999% CAS No.: 7440-50-8 Physical Form: ≈15.8g/m Odour: Odourless MDL No.: MFCD00010965
Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545