Zinc 99+%, mossy
Supplier: Thermo Scientific
ACRO201450050EA 299 EUR
ACRO201450050 ACRO201455000 ACRO201450010
Zinc 99+%, mossy
Zinc
Formula: Zn
MW: 65,39 g/mol
Boiling Pt: 907 °C (760 mmHg)
Melting Pt: 419 °C
Density: 7,14 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
Merck Index: 12,10255
Order Now
Specification Test Results
Appearance Silver grey Mossy flakes
Total trace metal impurities ≤1.0 %