Zinc 99,9 Metals basis, rods, Ø 1.27 cm (0.5 in)
Supplier: Thermo Fisher Scientific
00422.0R 00422.BT 00422.RM
00422.0REA 143 EUR
00422.0R 00422.BT 00422.RM
Zinc 99,9 Metals basis, rods, Ø 1.27 cm (0.5 in)
Zinc
Purity: 99.9+% CAS No.: 7440-66-6 Physical Form: ≈9.03g/cm Odour: Odourless MDL No.: MFCD00011291
Formula: Zn
MW: 65.39 g/mol
Boiling Pt: 908
Melting Pt: 420
Density: 7.14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255