Zinc ≥99.9% (metals basis), foil, Width: 30 cm (12 in), Thickness: 0.62 mm (0.024 in)
Supplier: Thermo Fisher Scientific
10437.CB 10437.BW
10437.CBEA 345 EUR
10437.CB 10437.BW
Zinc ≥99.9% (metals basis), foil, Width: 30 cm (12 in), Thickness: 0.62 mm (0.024 in)
Zinc
Purity: 99.9% CAS No.: 7440-66-6 Physical Form: ≈665g/30x50cm Odour: Odourless MDL No.: MFCD00011291
Formula: Zn
MW: 65.39 g/mol
Boiling Pt: 908
Melting Pt: 420
Density: 7.14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255