Zinc, powder, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
96454-1KG 96454-250G
SIAL96454-1KGEA 103 EUR
SIAL96454-1KG SIAL96454-250G
Zinc, powder, Sigma-Aldrich®
Zinc
Formula: Zn
MW: 65.39 g/mol
Boiling Pt: 908
Melting Pt: 420
Density: 7.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
UN: 1436
ADR: 4.3 II
Merck Index: 12,10255