Chromium ≥99.2% (metals basis), powder APS <10 µm
Supplier: Thermo Scientific
Warning
41797.18 41797.30 41797.A1
41797.18EA 86.1 EUR
41797.18 41797.30 41797.A1
Chromium ≥99.2% (metals basis), powder APS <10 µm
Chromium
Purity: 99.2% CAS No.: 7440-47-3 Odour: Odourless UN No.: 3089 MDL No.: MFCD00010944
Formula: Cr
MW: 52 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010944
CAS Number: 7440-47-3
EINECS: 231-157-5
UN: 3089
ADR: 4.1,III