Xylene (mixture of isomers) ≥98%, TECHNICAL
Supplier: VWR Chemicals
Danger
Synonyms: Dimethylbenzene
28973.294EA 67.1 EUR
28973.294 28973.328 28973.556 28973.363 28973.465 28973.950
Xylene (mixture of isomers) ≥98%, TECHNICAL
Xylene (mixture of isomers)
Formula: C₆H₄(CH₃)₂
Boiling Pt: 140 °C (1013 hPa)
Melting Pt: –34 °C
Density: 0.86 g/cm³ (20 °C)
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00077264
CAS Number: 1330-20-7
EINECS: 215-535-7
UN: 1307
ADR: 3,III
REACH: 01-2119488216-32
Merck Index: 13,10136
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2deefc7b7b67c10e64003e243ff3216fad27cf7d&vid=9fa01ddafd4814d5f34266ae7933463a29d92d19&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay Min. 98%