Xylene (mixture of isomers)

Supplier: Roth Carl
Danger

9713.1 9713.2 9713.3 9713.4
ROTH9713.1EA 33.9 EUR
ROTH9713.1 ROTH9713.2 ROTH9713.3 ROTH9713.4
Xylene (mixture of isomers)
Xylene (mixture of isomers)
Formula: C₆H₄(CH₃)₂
Boiling Pt: 140 °C (1013 hPa)
Melting Pt: –34 °C
Density: 0.86…0.88 g/cm³ (20 °C)
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00077264
CAS Number: 1330-20-7
EINECS: 215-535-7
UN: 1307
ADR: 3,III
Merck Index: 13,10136
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2deefc7b7b67c10e64003e243ff3216fad27cf7d&vid=9fa01ddafd4814d5f34266ae7933463a29d92d19&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR