Methanol-[D₁] 99.0 atom % D NMR
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Carbinol-D1, Methanol-[²H₁]
299130250 299131000
ACRO299130250EA 55.5 EUR
ACRO299130250 ACRO299131000
Methanol-[D₁] 99.0 atom % D NMR
Methanol-[D1]
Formula: CH₃DO
MW: 33.03 g/mol
Boiling Pt: 64.5
Melting Pt: –98 °C
Density: 0.81 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00044213
CAS Number: 1455-13-6
EINECS: 215-933-0
UN: 1230
ADR: 3 II
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Water ≤0.1 % (Coulometric)
D-Enrichment ≥99.0 %