Chlorobenzene-[D₅] 98.5+ atom % D for NMR spectroscopy
Supplier: Thermo Scientific
Warning
Synonyms: Chlorobenzene-[²H₅], Phenylchloride-D5
175320050 175320010
ACRO175320050EA 224 EUR
ACRO175320050 ACRO175320010
Chlorobenzene-[D₅] 98.5+ atom % D for NMR spectroscopy
Chlorobenzene-[D5]
Formula: C₆D₅Cl
MW: 117.52 g/mol
Boiling Pt: 130 °C (1013 hPa)
Melting Pt: –46 °C
Density: 1,15 g/cm³ (20 °C)
Flash Pt: 29 °C
Storage Temperature: Ambient
MDL Number: MFCD00000531
CAS Number: 3114-55-4
EINECS: 221-482-0
UN: 1134
ADR: 3,III
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Water ≤0.05 %
D-Enrichment ≥98.50 %
HPLC ≥97.5 %