Formalin 10% v/v, histology grade, neutralized (pH 7.0 ± 0.2), J.T.Baker®
Supplier: Avantor
Warning
Synonyms: Paraformaldehyde aqueous solution, Formaldehyde, Methylaldehyde, Formol
3933.9020EA 36.49 EUR
3933.9020 3933.5000PC 3933.9010 3933.1000 3933.1000MB 3933.9010JL 3933.9020JL
Formalin 10% v/v, histology grade, neutralized (pH 7.0 ± 0.2), J.T.Baker®
Formalin
Formula: CH₂O
MW: 30.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
EINECS: 200-001-8
IVDLogo
Order Now
Specification Test Results
Sodium (Na) 61 - 71 mmol/l
Formaldehyde (HCHO) 38 - 42 g/l
pH at 25°C 6.8 - 7.2