Ethylamine 70% ≥70% in aqueous solution
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Monoethylamine, Aminoethane, Ethanamine
A15022.0F A15022.AP
A15022.0FEA 83.1 EUR
A15022.0F A15022.AP
Ethylamine 70% ≥70% in aqueous solution
Ethylamine
Formula: C₂H₇N
MW: 45.08 g/mol
Boiling Pt: ∼38 °C
Melting Pt: –81 °C
Density: 0.803
Flash Pt: –17 °C (1 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008160
CAS Number: 75-04-7
EINECS: 200-834-7
UN: 2270
ADR: 3 II
Merck Index: 14,3762